Anti-GST Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB22845-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Monoclonal antibodies are produced by immunizing mouse with GST protein.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: