Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-FLAG peptide Mouse monoclonal antibody
 :  Biorbyt
 
Anti-FLAG peptide Mouse monoclonal antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB22842-0.5
  • BIRBORB22842-0.2
  • Antigen symbol:
  • FLAG
  • FLAG
  • Antigen name:
  • FLAG peptide
  • FLAG peptide
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Immunogen:
  • Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (DYKDDDDK), coupled to KLH.
  • Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (DYKDDDDK), coupled to KLH.
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together