Anti-FLAG peptide Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: FLAG peptide
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity:
Specifications
- Catalog No:
- BIRBORB22841-200
- BIRBORB22841-100
- Antigen symbol:
- FLAG
- FLAG
- Antigen name:
- FLAG peptide
- FLAG peptide
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (DYKDDDDK), coupled to KLH.
- Monoclonal antibodies are produced by immunizing mouse with synthetic peptide (DYKDDDDK), coupled to KLH.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together