Anti-C1Q Goat Polyclonal Antibody
BIRBORB21929-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:C1Q
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Format:antiserum
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: C1Q
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: