Anti-IgG Polyclonal Antibody (Biotin)
BIRBORB21629-1
: Biorbyt
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Biotin/IgG protein molar ratio (B/P) is approximately 5,4, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG isolated from pooled human serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype: none
Reactivity: