Anti-MAP2K4 Rabbit Polyclonal Antibody
Catalog # BIRBORB14887-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 4
- Antigen symbol:MAP2K4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Modification:phospho-Thr261
- Storage buffer:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human SEK1/MKK4 around the phosphorylation site of threonine261 (A -K-TP-R-D).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunochemistry: 1:50-1:100 Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat