Anti-PRKAA1 Rabbit Polyclonal Antibody
BIRBORB14885-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Modification:phospho-Ser487
- Storage buffer:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human AMPK alpha1around the phosphorylation site of serine 487 (S -G- SP-V-S).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100 Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat