Anti-Neuropilin and tolloid-like protein 1 Mouse Monoclonal Antibody
Catalog # BIRBORB13213-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Neuropilin and tolloid-like Protein 1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Storage buffer:TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
- Molecular weight:47 kDa
- Storage temperature:Cool
- Concentration:0,3 mg/ml
- Shipping temperature:Ice
- Immunogen:This antibody is produced by immunizing mouse with recombinant protein NETO1.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:1000-2000; Immunofluorescence: 1:100-500
Type: Primary
Antigen: Neuropilin and tolloid-like protein 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse