Anti-MEI-41 Goat Polyclonal Antibody
Catalog # BIRBORB12347-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:MEI-41
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Drosophila
- Storage buffer:0,02% sodium azide, pH 7,3 with 0,5% BSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MEI-41
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Drosophila