Anti-RAET1E Mouse Monoclonal Antibody [clone: 2D11]
Catalog # ABNOH00135250-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Retinoic Acid Early Transcript 1, epsilon
- Antigen symbol:RAET1E
- Clonality:Monoclonal
- Clone:2D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:135250
- Antigen synonyms:ULBP4|RL-4|RAET1E2|bA350J20.7|NKG2DL4|LETAL|N2DL-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RAET1E
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human