Anti-FRA10AC1 Mouse Polyclonal Antibody
ABNOH00118924-A01
: Abnova
- Antibody type:Primary
- Antigen name:fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
- Antigen symbol:FRA10AC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:118924
- Antigen synonyms:FRA10A|C10orf4|F26C11.1-like
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: FRA10AC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human