Anti-CYGB Mouse Monoclonal Antibody [clone: 3A5-2D2]
Catalog # ABNOH00114757-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytoglobin B
- Antigen symbol:CYGB
- Clonality:Monoclonal
- Clone:3A5-2D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:114757
- Antigen synonyms:STAP|HGB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CYGB
Clonality: Monoclonal
Clone: 3A5-2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human