Anti-CEACAM21 Rabbit Polyclonal Antibody
ABNOH00090273-D01
: Abnova
- Antibody type:Primary
- Antigen name:Carcinoembryonic antigen-related cell adhesion molecule 21
- Antigen symbol:CEACAM21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:90273
- Antigen synonyms:R29124_1|CEACAM3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: CEACAM21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human