Anti-SIGLEC12 Mouse Monoclonal Antibody [clone: 1D1]
Catalog # ABNOH00089858-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid-binding Ig-like lectin 12
- Antigen symbol:SIGLEC12
- Clonality:Monoclonal
- Clone:1D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:89858
- Antigen synonyms:mSiglec-E|Siglec9|SIGLECL1|Siglec5|Siglec12|Cd170|Siglecl1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SIGLEC12
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human