Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SIGLEC10 Mouse Monoclonal Antibody [clone: 1D11]
Anti-SIGLEC10 Mouse Monoclonal Antibody [clone: 1D11]
  ABNOH00089790-M01
 :  Abnova
undefined
Anti-SIGLEC10 Mouse Monoclonal Antibody [clone: 1D11]
  ABNOH00089790-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Sialic acid binding Ig-like lectin 10
  • Antigen symbol:
    SIGLEC10
  • Clonality:
    Monoclonal
  • Clone:
    1D11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    89790
  • Antigen synonyms:
    PRO940|SLG2|SIGLEC-10
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: SIGLEC10
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human