Anti-TSSK6 Rabbit Polyclonal Antibody
Catalog # ABNOH00083983-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Testis-specific serine kinase 6
- Antigen symbol:TSSK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:83983
- Antigen synonyms:CT72|TSSK4|FKSG82|SSTK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TSSK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human