Anti-DBF4B Mouse Monoclonal Antibody [clone: 1A7]
Catalog # ABNOH00080174-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DBF4 Zinc Finger B
- Antigen symbol:DBF4B
- Clonality:Monoclonal
- Clone:1A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80174
- Antigen synonyms:ASKL1, CHIFB, DRF1, ZDBF1B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DBF4B
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human