Anti-ASRGL1 Mouse Polyclonal Antibody
ABNOH00080150-A01
: Abnova
- Antibody type:Primary
- Antigen name:Asparaginase Like 1
- Antigen symbol:ASRGL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80150
- Antigen synonyms:ALP1|ASRGL1|ALP|FLJ22316|CRASH|asparaginase like 1|asparaginase-like 1 protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: ASRGL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human