Anti-ZNF71 Mouse Monoclonal Antibody [clone: 3F4]
ABNOH00058491-M01
: Abnova
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 71
- Antigen symbol:ZNF71
- Clonality:Monoclonal
- Clone:3F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:58491
- Antigen synonyms:EZFIT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZNF71
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human