Anti-CSAD Mouse Monoclonal Antibody [clone: 1D2]
Catalog # ABNOH00051380-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cysteine sulfinic acid decarboxylase
- Antigen symbol:CSAD
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51380
- Antigen synonyms:CSD|PCAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: CSAD
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human