Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: 3H1]
Catalog # ABNOH00027036-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 7
- Antigen symbol:SIGLEC7
- Clonality:Monoclonal
- Clone:3H1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:27036
- Antigen synonyms:CD328|SIGLECP2|p75/AIRM1|p75|CDw328|D-siglec|SIGLEC19P|AIRM1|QA79|SIGLEC-7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human