Anti-ICOSL Mouse Monoclonal Antibody [clone: 2C2]
Catalog # ABNOH00023308-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Inducible T-cell Co-stimulator Ligand
- Antigen symbol:ICOSL
- Clonality:Monoclonal
- Clone:2C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23308
- Antigen synonyms:ICOS-L|B7-H2|B7RP-1|CD275|LICOS|B7RP1|ICOSL|GL50|B7H2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ICOSL
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human