Anti-EXOSC7 Mouse Polyclonal Antibody
Catalog # ABNOH00023016-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Exosome Component 7
- Antigen symbol:EXOSC7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23016
- Antigen synonyms:Rrp42p|p8|RRP42|EAP1|hRrp42p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: EXOSC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human