Anti-CORO1A Mouse Monoclonal Antibody [clone: 4G10]
Catalog # ABNOH00011151-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Coronin 1A
- Antigen symbol:CORO1A
- Clonality:Monoclonal
- Clone:4G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11151
- Antigen synonyms:HCORO1|IMD8|p57|CLABP|CLIPINA|TACO
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CORO1A
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human