Anti-ADAMTS8 Mouse Monoclonal Antibody [clone: 5A3]
ABNOH00011095-M01
: Abnova
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 8
- Antigen symbol:ADAMTS8
- Clonality:Monoclonal
- Clone:5A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11095
- Antigen synonyms:METH2|ADAM-TS8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ADAMTS8
Clonality: Monoclonal
Clone: 5A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human