Anti-LIAS Mouse Monoclonal Antibody [clone: 1C7]
Catalog # ABNOH00011019-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lipoic Acid Synthetase
- Antigen symbol:LIAS
- Clonality:Monoclonal
- Clone:1C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11019
- Antigen synonyms:HUSSY-01|PDHLD|LS|LAS|LIP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LIAS
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human