Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: 2H5]
Catalog # ABNOH00010241-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Calcium Binding And Coiled-coil Domain 2
- Antigen symbol:CALCOCO2
- Clonality:Monoclonal
- Clone:2H5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10241
- Antigen synonyms:Calcium binding and coiled-coil domain 2|NDP52
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CALCOCO2
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human