Anti-NOLC1 Mouse Polyclonal Antibody
Catalog # ABNOH00009221-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nucleolar and coiled-body phosphoprotein 1
- Antigen symbol:NOLC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9221
- Antigen synonyms:NOPP130|nucleolar and coiled-body phosphoprotein 1|NOPP140|KIAA0035|P130|nucleolar phosphoprotein p130|nucleolar protein p130|HCV NS5A trans-regulated protein 13|nucleolar and coiled-body phosphprotein 1|NS5ATP13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: NOLC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human