Order Entry
United Kingdom
ContactUsLinkComponent
Anti-RNMT Mouse Monoclonal Antibody [clone: 3H3-1D12]
Anti-RNMT Mouse Monoclonal Antibody [clone: 3H3-1D12]
Catalog # ABNOH00008731-M01
Supplier:  Abnova
undefined
Anti-RNMT Mouse Monoclonal Antibody [clone: 3H3-1D12]
Catalog # ABNOH00008731-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RNA (guanine-7-) methyltransferase
  • Antigen symbol:
    RNMT
  • Clonality:
    Monoclonal
  • Clone:
    3H3-1D12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    8731
  • Antigen synonyms:
    cm1p|Met|hCMT1|RG7MT1|hMet|CMT1|CMT1c|MET
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: RNMT
Clonality: Monoclonal
Clone: 3H3-1D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human