Anti-ANP32A Mouse Monoclonal Antibody [clone: 2G11-4A5]
Catalog # ABNOH00008125-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Acidic Nuclear Phosphoprotein 32 Family, Member A
- Antigen symbol:ANP32A
- Clonality:Monoclonal
- Clone:2G11-4A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8125
- Antigen synonyms:C15orf1|I1PP2A|PHAPI|LANP|HPPCn|MAPM|PP32|PHAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ANP32A
Clonality: Monoclonal
Clone: 2G11-4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human