Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-TAF1 Mouse Monoclonal Antibody [clone: 1E11]
  ABNOH00006872-M01
 :  Abnova
undefined
Anti-TAF1 Mouse Monoclonal Antibody [clone: 1E11]
  ABNOH00006872-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa
  • Antigen symbol:
    TAF1
  • Clonality:
    Monoclonal
  • Clone:
    1E11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6872
  • Antigen synonyms:
    DYT3|TAF2A|DYT3/TAF1|P250|BA2R|CCGS|N-TAF1|NSCL2|KAT4|TAFII-250|CCG1|OF|TAFII250|TAF(II)250|XDP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: TAF1
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human