Anti-SPARC Mouse Monoclonal Antibody [clone: 1B2]
Catalog # ABNOH00006678-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Secreted Protein, Acidic, Cysteine-Rich
- Antigen symbol:SPARC
- Clonality:Monoclonal
- Clone:1B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6678
- Antigen synonyms:Cysteine-Rich Protein|Secreted Protein Acidic And Rich In Cysteine|ON|Basement-Membrane Protein 40|Osteonectin|BM-40
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human