Anti-RARA Mouse Monoclonal Antibody [clone: 2C3]
ABNOH00005914-M09
: Abnova
- Antibody type:Primary
- Antigen name:Retinoic Acid Receptor Alpha
- Antigen symbol:RARA
- Clonality:Monoclonal
- Clone:2C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5914
- Antigen synonyms:Retinoic acid receptor|nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form|NR1B1|alpha polypeptide|RAR|nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR short form
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RARA
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human