Order Entry
United Kingdom
ContactUsLinkComponent
Anti-NDUFV1 Mouse Monoclonal Antibody [clone: 4A7]
Anti-NDUFV1 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00004723-M01
Supplier:  Abnova
undefined
Anti-NDUFV1 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00004723-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa
  • Antigen symbol:
    NDUFV1
  • Clonality:
    Monoclonal
  • Clone:
    4A7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    4723
  • Antigen synonyms:
    CI-51K|UQOR1|CI51KD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: NDUFV1
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human