Anti-MARS Mouse Monoclonal Antibody [clone: 5G5]
Catalog # ABNOH00004141-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Methionyl tRNA synthetase
- Antigen symbol:MARS
- Clonality:Monoclonal
- Clone:5G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4141
- Antigen synonyms:MRS|SPG70|MTRNS|CMT2U|METRS|ILLD|ILFS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MARS
Clonality: Monoclonal
Clone: 5G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human