Anti-FADS1 Mouse Monoclonal Antibody [clone: 2D9]
Catalog # ABNOH00003992-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Desaturase 1
- Antigen symbol:FADS1
- Clonality:Monoclonal
- Clone:2D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3992
- Antigen synonyms:FADSD5|TU12 |FADS1|fatty acid desaturase 1 |delta-5 fatty acid desaturase|FLJ90273|linoleoyl-CoA desaturase (delta-6-desaturase)-like 1|LLCDL1|delta-5 desaturase|FADS6|D5D|FLJ38956
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FADS1
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human