Anti-LIPA Mouse Monoclonal Antibody [clone: 1F9]
Catalog # ABNOH00003988-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lipase A, Lysosomal Acid, Cholesterol Esterase
- Antigen symbol:LIPA
- Clonality:Monoclonal
- Clone:1F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3988
- Antigen synonyms:CESD|LAL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LIPA
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human