Anti-ISL1 Mouse Monoclonal Antibody [clone: 5F12]
Catalog # ABNOH00003670-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Islet 1
- Antigen symbol:ISL1
- Clonality:Monoclonal
- Clone:5F12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3670
- Antigen synonyms:Isl-1|ISLET1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ISL1
Clonality: Monoclonal
Clone: 5F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human