Anti-PDX1 Mouse Monoclonal Antibody [clone: 4E12]
Catalog # ABNOH00003651-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pancreatic and duodenal homeobox 1
- Antigen symbol:PDX1
- Clonality:Monoclonal
- Clone:4E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3651
- Antigen synonyms:Islet/duodenum homeobox-1|IPF-1|Insulin promoter factor 1|Pancreas/duodenum homeobox protein 1|IPF1|Somatostatin-transactivating factor 1|Insulin upstream factor 1|IUF-1|PDX-1|GSF|STF-1|Glucose-sensitive factor|PDX1|IDX-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PDX1
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human