Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HMGCS2 Mouse Monoclonal Antibody [clone: 1E9]
Anti-HMGCS2 Mouse Monoclonal Antibody [clone: 1E9]
Catalog # ABNOH00003158-M06
Supplier:  Abnova
undefined
Anti-HMGCS2 Mouse Monoclonal Antibody [clone: 1E9]
Catalog # ABNOH00003158-M06
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3-hydroxy-3-methylglutaryl-CoA Synthase 2 (mitochondrial)
  • Antigen symbol:
    HMGCS2
  • Clonality:
    Monoclonal
  • Clone:
    1E9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    3158
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: HMGCS2
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human