Anti-GBA Mouse Monoclonal Antibody [clone: 2E2]
Catalog # ABNOH00002629-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glucosidase, Beta, Acid
- Antigen symbol:GBA
- Clonality:Monoclonal
- Clone:2E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2629
- Antigen synonyms:GBA1|GLUC|GCB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GBA
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human