Anti-FASN Mouse Monoclonal Antibody [clone: 3A2]
Catalog # ABNOH00002194-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Synthase
- Antigen symbol:FAS
- Clonality:Monoclonal
- Clone:3A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:2194
- Antigen synonyms:FAS|SDR27X1|OA-519
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FASN
Clonality: Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human