Anti-FASN Mouse Monoclonal Antibody [clone: 3F2-1F3]
Catalog # ABNOH00002194-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Synthase
- Antigen symbol:FAS
- Clonality:Monoclonal
- Clone:3F2-1F3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2194
- Antigen synonyms:FAS|SDR27X1|OA-519
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: FASN
Clonality: Monoclonal
Clone: 3F2-1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human