Anti-FABP2 Mouse Monoclonal Antibody [clone: 2F7]
Catalog # ABNOH00002169-M11
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 2, Intestinal
- Antigen symbol:FABP2
- Clonality:Monoclonal
- Clone:2F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:2169
- Antigen synonyms:I-FABP|FABPI
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FABP2
Clonality: Monoclonal
Clone: 2F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human