Anti-FABP1 Mouse Monoclonal Antibody [clone: 5F7]
Catalog # ABNOH00002168-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 1
- Antigen symbol:FABP1
- Clonality:Monoclonal
- Clone:5F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:2168
- Antigen synonyms:SCP|FABPL|p14|SCP.|L-FABP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: FABP1
Clonality: Monoclonal
Clone: 5F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human