Anti-FABP4 Mouse Monoclonal Antibody [clone: 2F12-1B11]
Catalog # ABNOH00002167-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 4
- Antigen symbol:FABP4
- Clonality:Monoclonal
- Clone:2F12-1B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:2167
- Antigen synonyms:ALBP|A-FABP|adipocyte-type fatty acid-binding protein|fatty acid-binding protein 4|AFABP|aP2|HEL-S-104
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: FABP4
Clonality: Monoclonal
Clone: 2F12-1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human