Anti-EXT1 Mouse Monoclonal Antibody [clone: 5A5]
Catalog # ABNOH00002131-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Exostosin 1
- Antigen symbol:EXT1
- Clonality:Monoclonal
- Clone:5A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2131
- Antigen synonyms:TTV|LGS|TRPS2|EXT|LGCR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EXT1
Clonality: Monoclonal
Clone: 5A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human