Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CRX Mouse Monoclonal Antibody [clone: 6D11]
Anti-CRX Mouse Monoclonal Antibody [clone: 6D11]
Catalog # ABNOH00001406-M04
Supplier:  Abnova
undefined
Anti-CRX Mouse Monoclonal Antibody [clone: 6D11]
Catalog # ABNOH00001406-M04
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cone-rod Homeobox
  • Antigen symbol:
    CRX
  • Clonality:
    Monoclonal
  • Clone:
    6D11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1406
  • Antigen synonyms:
    LCA7|OTX3|CRD|CORD2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CRX
Clonality: Monoclonal
Clone: 6D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat