Anti-FAS Mouse Monoclonal Antibody [clone: 1B6]
Catalog # ABNOH00000355-M11
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Synthase
- Antigen symbol:FAS
- Clonality:Monoclonal
- Clone:1B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:355
- Antigen synonyms:FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FAS
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human