Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ADD1 Mouse Monoclonal Antibody [clone: 2C9]
  ABNOH00000118-M01
 :  Abnova
undefined
Anti-ADD1 Mouse Monoclonal Antibody [clone: 2C9]
  ABNOH00000118-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Adducin 1 Alpha
  • Antigen symbol:
    ADD1
  • Clonality:
    Monoclonal
  • Clone:
    2C9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    118
  • Antigen synonyms:
    ADDA|ADD2|Adducin-alpha|alpha-Adducin|Adducin1|Adducin-1|Erythrocyte adducin subunit beta|beta-Adducin|beta Adducin|Beta-adducin|Adducin beta|ADD1|Erythrocyte adducin subunit alpha|Adducin-beta|alpha Adducin|Alpha-adducin|ADDB
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ADD1
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human